Anti-CD99 Mouse Monoclonal Antibody [clone: 3B2/TA8] (APC (Allophycocyanin))
ANTIA85604-100
New Product
- Antibody type:Primary
- Antigen symbol:CD99
- Clonality:Monoclonal
- Clone:3B2/TA8
- Conjugation:APC (Allophycocyanin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Seq:0790
- Form:Liquid
- Gene ID:UniprotID# P14209
- Antigen synonyms:12E7|CD99 antigen|E2 antigen|MIC2|MIC2X|MIC2Y|Protein MIC2|T-cell surface glycoprotein E2
- Storage buffer:Supplied in phosphate buffered saline, pH 7,4, with 15 mM sodium azide
- Storage temperature:Store in the dark at 2...8 °C. Do not freeze.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Human thymocytes
- Purification:This antibody is conjugated with Allophycocyanin (APC) under optimum conditions. The conjugate is purified by size-exclusion chromatography; removing unconjugated antibody and free fluorochrome.
- Pack type:Glass bottle
Mouse monoclonal [3B2/TA8] antibody to CD99 (APC) for Flow Cytometry with samples derived from Human.
- Recommended dilutions: Flow Cytometry: The reagent is designed for analysis of human blood cells using 10 µl reagent / 100 µl of whole blood or 106 cells in a suspension
- The content of a vial (1 ml) is sufficient for 100 tests
Type: Primary
Antigen: CD99
Clonality: Monoclonal
Clone: 3B2/TA8
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human