Order Entry
Export
ContactUsLinkComponent
Bovine Insulin ELISA Kit (High Sensitivity)
Bovine Insulin ELISA Kit (High Sensitivity)
Catalog # ANTIA319378-96
Supplier:  ANTIBODIES.COM
New Product
CAS Number:  
Bovine Insulin ELISA Kit (High Sensitivity)
Catalog # ANTIA319378-96
Supplier:  ANTIBODIES.COM
New Product
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Bovine
  • Target protein:
    Insulin
  • Description:
    Bovine Insulin ELISA kit (High Sensitivity)
  • Sample type:
    Serum, plasma, tissue homogenates, and other biological fluids
  • Cross reactivity:
    Bovine Insulin ELISA Kit (High Sensitivity) exhibits high specificity and excellent specificity for the detection of bovine Insulin. No significant cross-reactivity or interference between Insulin and analogues was observed.
  • Detection method:
    Colorimetric
  • Time to Results:
    4 hours
  • Shelf life:
    Store for 6 months at 4 °C
  • Detection range:
    7,813 - 500 pg/ml
  • Storage temperature:
    4 °C
  • Sample volume:
    100 μl
  • Sensitivity:
    4,688 pg/ml
  • Regulatory status:
    RUO
  • Tests per kit:
    96 tests

Specifications

About this item

Bovine Insulin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of bovine Insulin in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready-to-use ELISA kit
  • Assay Precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%

Bovine Insulin ELISA Kit (A319378) employs the sandwich enzyme immunoassay technique for the quantitative measurement of bovine Insulin in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Insulin has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Insulin present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Insulin Antibody, which binds the captured Insulin present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Insulin captured in each well. The concentration of Insulin can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Delivery: This is a Ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch and protocol manual.
Caution: This product is for Research use only. It is not for use in diagnostic or therapeutic procedures.