Order Entry
Export
ContactUsLinkComponent
Human IL-17A ELISA Kit (High Sensitivity)
Human IL-17A ELISA Kit (High Sensitivity)
Catalog # ANTIA323173-96
Supplier:  Antibodies.com
New Product
CAS Number:  
Human IL-17A ELISA Kit (High Sensitivity)
Catalog # ANTIA323173-96
Supplier:  Antibodies.com
New Product
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Human
  • Target protein:
    IL-17A
  • Description:
    Human IL-17A ELISA kit (high sensitivity)
  • Sample type:
    Serum, plasma, and other biological fluids
  • Cross reactivity:
    No significant cross-reactivity or interference between human IL-17A and analogues was observed.
  • Detection method:
    Colorimetric
  • Time to Results:
    3 h 30 min
  • Shelf life:
    Store for 6 months at 4 °C
  • Detection range:
    0,78 - 50 pg/ml
  • Storage temperature:
    4 °C
  • Sensitivity:
    0,49 pg/ml
  • Regulatory status:
    RUO
  • Tests per kit:
    96 tests

Specifications

About this item

Human IL-17A ELISA kit is a high sensitivity sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human IL-17A in serum, plasma, and other biological fluids.

  • High Sensitivity: Detection at the picogram level!
  • Assay Precision: Intra-Assay: CV <10%, Inter-Assay: CV <10%

Human IL-17A ELISA kit employs the sandwich Enzyme-Linked Immunoassay technique for the quantitative measurement of human IL-17A in serum, plasma, and other biological fluids. An antibody specific for IL-17A has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the IL-17A present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-IL-17A Antibody, which binds the captured IL-17A present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Avidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of IL-17A captured in each well. The concentration of IL-17A can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Caution: This product is for Research Use Only. It is not for use in diagnostic or therapeutic procedures.