Order Entry
Export
ContactUsLinkComponent
Human Recombinant CRCP (from HEK293 Cells)
Human Recombinant CRCP (from HEK293 Cells)
Supplier:  Antibodies.com
New Product
Certificates
Human Recombinant CRCP (from HEK293 Cells)
Supplier:  Antibodies.com
New Product

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage conditions:
    Lyophilised: Store at –20...–80 °C. Reconstituted: Aliquot and store at –80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
  • Gene ID:
    UniprotID# O75575
  • Reconstitution instructions:
    Reconstitute with distilled sterile water.
  • Carrier-free:
    N
  • Protein synonyms:
    Calcitonin gene-related peptide-receptor component protein|CGRP-RCP|CGRP-receptor component protein|CGRPRCP|DNA-directed RNA polymerase III subunit RPC9|HsC17|RNA polymerase III subunit C9
  • Protein/peptide name:
    CRCP
  • Purity:
    >95% by SDS-PAGE and Coomassie blue staining
  • Molecular weight:
    The protein has a predicted molecular mass of 43,0 kDa after removal of the signal peptide. The apparent molecular mass of hFc-CRCP is approximately 35 - 70 kDa due to glycosylation.
  • Sequence:
    hFc (Glu99-Ala330) + CRCP (Met1-Ala148)
  • Concentration:
    Reconstitution dependent
  • Formulation:
    Lyophilised from sterile Phosphate Buffered Saline, pH 7,4. Normally 5 - 8% trehalose is added as protectants before lyophilisation.
  • Shipping temperature:
    Shipped on blue ice at 4 °C.
  • Tested applications:
    SDS-PAGE

Specifications