Mouse Recombinant IL-11RA (from HEK293 Cells)
Supplier: Antibodies.com
New Product
Certificates
About this item
Partial length mouse IL-11RA, expressed in HEK293 cells.
Caution: This product is for research use only. It is not for use in diagnostic or therapeutic procedures.
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Storage conditions:Lyophilised: Store at –20...–80 °C. Reconstituted: Aliquot and store at –80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Gene ID:UniprotID# Q64385
- Reconstitution instructions:Reconstitute with distilled sterile water
- Carrier-free:N
- Protein synonyms:IL-11 receptor subunit alpha|IL-11R subunit alpha|IL-11R-alpha|IL11RA|Interleukin-11 receptor subunit alpha
- Protein/peptide name:IL-11RA
- Purity:>85% by SDS-PAGE and Coomassie blue staining
- Molecular weight:The protein has a predicted molecular mass of 38,3 kDa after removal of the signal peptide. The apparent molecular mass of mIL11RA-His is approximately 35 - 55 kDa due to glycosylation.
- Sequence:IL11RA (Cys26-Gln367) + 6×His Tag
- Concentration:Reconstitution dependent
- Formulation:Lyophilised from sterile phosphate buffered saline, pH 7,4. Normally 5 - 8% Trehalose is added as a protectant before lyophilisation.
- Shipping temperature:Shipped on blue ice at 4 °C
- Tested applications:SDS-PAGE