Human Recombinant CELA3B
Catalog # ANTIA251202-100
Supplier: Antibodies.com
New Product
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Species:Human
- Seq:1286
- Storage conditions:Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Gene ID:UniprotID# P08861
- Carrier-free:N
- Protein synonyms:Chymotrypsin-like elastase family member 3B;ELA3B;Elastase IIIB;Elastase-3B;Protease E
- Protein/peptide name:CELA3B
- Molecular weight:~17 kDa
- Concentration:100 µg/ml
- Formulation:Supplied in 10 mM Phosphate Buffered Saline with 0,05% Sodium Azide
- Shipping temperature:Shipped on blue ice at 4 °C
- Tested applications:IF, WB, IHC
Specifications
About this item
Human CELA3B suitable for IF, WB, IHC.
Caution: This product is for Research Use Only. It is not for use in diagnostic or therapeutic procedures.