Human Recombinant IL-36R (from HEK293 cells)
Supplier: Antibodies.com
New Product
Certificates
About this item
Partial length Human IL-36R, expressed in HEK293 cells.
Caution: This product is for Research Use Only. It is not for use in diagnostic or therapeutic procedures.
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:Lyophilised: Store at −20...−80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Gene ID:UniprotID# Q9HB29
- Reconstitution instructions:Reconstitute with distilled sterile water
- Carrier-free:N
- Protein synonyms:IL-1Rrp2|IL-36 receptor|IL-36R|IL1R-rp2|IL1RRP2|Interleukin-1 receptor-like 2|Interleukin-1 receptor-related protein 2
- Protein/peptide name:IL-36R
- Purity:>85% by SDS-PAGE and coomassie blue staining
- Molecular weight:The protein has a predicted molecular mass of 36,8 kDa after removal of the signal peptide
- Sequence:IL1RL2 (Asp20-Arg335) + 6×His Tag
- Concentration:Reconstitution dependent
- Formulation:Lyophilised from sterile Phosphate Buffered Saline, pH 7,4. Normally 5 - 8% Trehalose is added as a protectant before lyophilisation.
- Tested applications:SDS-PAGE