Order Entry
Export
ContactUsLinkComponent
Dog Recombinant PD1 (from HEK293 Cells)
Dog Recombinant PD1 (from HEK293 Cells)
Supplier:  Antibodies.com
New Product
Certificates
Dog Recombinant PD1 (from HEK293 Cells)
Supplier:  Antibodies.com
New Product

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Dog
  • Storage conditions:
    Lyophilised: Store at −20...−80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
  • Gene ID:
    UniprotID# A0A090BAM7
  • Reconstitution instructions:
    Reconstitute with distilled sterile water
  • Carrier-free:
    N
  • Protein synonyms:
    CD279|PDCD1|Programmed cell death protein 1|Protein PD-1
  • Protein/peptide name:
    PD1
  • Purity:
    >95% by SDS-PAGE and Coomassie blue staining
  • Molecular weight:
    The protein has a predicted molecular mass of 42,4 kDa after removal of the signal peptide. The apparent molecular mass of dPD-1-mFc is approximately 55 - 70 kDa due to glycosylation.
  • Sequence:
    PD-1 (Leu25-Gly168) + mFc (Pro99-Lys330)
  • Concentration:
    Reconstitution dependent
  • Formulation:
    Lyophilised from sterile phosphate buffered saline, pH 7,4. Normally 5 - 8% trehalose is added as a protectant before lyophilisation.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Tested applications:
    SDS-PAGE, ELISA, Flow Cytometry

Specifications