Order Entry
Export
ContactUsLinkComponent
Human Recombinant LRP5 (from HEK293 Cells)
Human Recombinant LRP5 (from HEK293 Cells)
Supplier:  Antibodies.com
New Product
Certificates
Human Recombinant LRP5 (from HEK293 Cells)
Supplier:  Antibodies.com
New Product

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage conditions:
    Shipped at 4 °C. Lyophilised: Store at −20...−80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
  • Gene ID:
    UniprotID# O75197
  • Reconstitution instructions:
    Reconstitute with distilled sterile water.
  • Carrier-free:
    N
  • Protein synonyms:
    Low-density lipoprotein receptor-related protein 5|Low-density lipoprotein receptor-related protein 7|LR3|LRP-5|LRP-7|LRP7
  • Protein/peptide name:
    LRP5
  • Purity:
    >95% by SDS-PAGE and Coomassie blue staining
  • Molecular weight:
    The protein has a predicted molecular mass of 95,9 kDa after removal of the signal peptide.
  • Sequence:
    hFc (Glu99-Ala330) + LRP5 (Glu644-Gln1263)
  • Concentration:
    Reconstitution dependent
  • Formulation:
    Lyophilised from sterile Phosphate Buffered Saline, pH 7,4. Normally 5 - 8% trehalose is added as protectants before lyophilisation.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Tested applications:
    SDS-PAGE

Specifications