Mouse Recombinant SLAMF7/CS1 (from HEK293 Cells)
New Product
Partial length Mouse SLAMF7/CS1, expressed in HEK293 cells.
: This product is for Research Use Only. It is not for use in diagnostic or therapeutic procedures.
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Storage conditions:Lyophilised: Store at –20...–80 °C. Reconstituted: Aliquot and store at –80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Gene ID:UniprotID# Q8BHK6
- Reconstitution instructions:Prior to reconstitution, centrifuge the vial at 5000 g for 3 - 5 minutes at room temperature. Reconstitute with appropriate volume of distilled sterile water to bring product to 1 mg/ml concentration. After addition of distilled sterile water, mix by gentle tapping. Note: It is not recommended to vortex or vigorously pipette the sample.
- Carrier-free:N
- Protein/peptide name:SLAMF7/CS1
- Purity:>95% by SDS-PAGE and Coomassie blue staining
- Molecular weight:The protein has a predicted molecular mass of 48,4 kDa after removal of the signal peptide. The apparent molecular mass of mCS1-hFc is approximately 55 - 70 due to glycosylation.
- Sequence:CS1 (Ser23-Gly224) + hFc (Glu99-Ala330)
- Concentration:Reconstitution dependent
- Formulation:Lyophilised from sterile Phosphate Buffered Saline, pH 7,4. Normally 5 - 8% trehalose is added as a protectant before lyophilisation.
- Shipping temperature:Shipped on blue ice at 4 °C.
- Tested applications:SDS-PAGE