Cynomolgus macaque Recombinant DLL3 (from HEK293 Cells)
New Product
Partial length Cynomolgus macaque DLL3, expressed in HEK293 cells.
: This product is for Research Use Only. It is not for use in diagnostic or therapeutic procedures.
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Cynomolgus macaque
- Storage conditions:Lyophilised: Store at −20...−80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Gene ID:UniprotID# A0A2K5WSR1
- Reconstitution instructions:Reconstitute with distilled sterile water
- Carrier-free:N
- Protein synonyms:Delta-like protein 3|Delta3|Drosophila Delta homolog 3
- Protein/peptide name:DLL3
- Purity:>95% by SDS-PAGE and Coomassie blue staining
- Molecular weight:The protein has a predicted molecular mass of 44,5 kDa after removal of the signal peptide. The apparent molecular mass of cDLL3(309-488)-hFc is approximately 35 - 70 kDa due to glycosylation.
- Sequence:DLL3 (Val309-Arg488) + hFc (Glu99-Ala330)
- Concentration:Reconstitution dependent
- Formulation:Lyophilised from sterile Phosphate Buffered Saline, pH 7,4. Normally 5 - 8% trehalose is added as protectants before lyophilization.
- Shipping temperature:Shipped on blue ice at 4 °C
- Tested applications:SDS-PAGE