Human Recombinant CD11b (from HEK293 Cells)
New Product
Partial length Human CD11b, expressed in HEK293 cells.
: This product is for esearch use only. It is not for use in diagnostic or therapeutic procedures.
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:Lyophilised: Store at −20...−80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Gene ID:UniprotID# P11215
- Reconstitution instructions:Prior to reconstitution, centrifuge the vial at 5000 g for 3 - 5 minutes at room temperature. Reconstitute with appropriate volume of distilled sterile water to bring product to 1 mg/ml concentration. After addition of distilled sterile water, mix by gentle tapping. Note: It is not recommended to vortex or vigorously pipette the sample.
- Carrier-free:N
- Protein synonyms:CD11 antigen-like family member B|Cell surface glycoprotein MAC-1 subunit alpha|CR-3 alpha chain|CR3A|Integrin alpha-M|ITGAM|Leukocyte adhesion receptor MO1|Neutrophil adherence receptor
- Protein/peptide name:CD11b
- Purity:>95% by SDS-PAGE and Coomassie blue staining
- Molecular weight:The protein has a predicted molecular mass of 145,2 kDa after removal of the signal peptide
- Sequence:ITGAM (Phe17-Asn1104) + hFc (Glu99-Ala330)
- Concentration:Reconstitution dependent
- Formulation:Lyophilised from sterile phosphate buffered saline, pH 7,4. Normally 5 - 8% trehalose is added as a protectant before lyophilisation.
- Shipping temperature:Shipped on blue ice at 4 °C