Human Recombinant MUC1 (from HEK293 Cells)
New Product
Partial length Human MUC1, expressed in HEK293 cells.
: This product is for Research Use Only. It is not for use in diagnostic or therapeutic procedures.
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:Lyophilised: Store at −20...−80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Gene ID:UniprotID# P15941
- Reconstitution instructions:Prior to reconstitution, centrifuge the vial at 5000 g for 3 - 5 minutes at room temperature. Reconstitute with appropriate volume of distilled sterile water to bring product to 1 mg/ml concentration. After addition of distilled sterile water, mix by gentle tapping. Note: It is not recommended to vortex or vigorously pipette the sample.
- Carrier-free:N
- Protein synonyms:Breast carcinoma-associated antigen DF3|CA 15-3|Cancer antigen 15-3|Carcinoma-associated mucin|CD227|Episialin|H23AG|MUC-1|Mucin-1|PUM
- Protein/peptide name:MUC1
- Purity:>95% by SDS-PAGE and Coomassie blue staining
- Molecular weight:The protein has a predicted molecular mass of 39,7 kDa after removal of the signal peptide
- Sequence:MUC1 (Leu1036-Val1156) + hFc (Glu99-Ala330)
- Concentration:Reconstitution dependent
- Formulation:Lyophilised from sterile Phosphate Buffered Saline, pH 7,4. Normally 5 - 8% Trehalose is added as a protectant before lyophilization.
- Shipping temperature:Shipped on blue ice at 4 °C
- Tested applications:SDS-PAGE