Human Recombinant CDKN2A/p14ARF (from HEK293 Cells)
Supplier: Antibodies.com
New Product
Certificates
About this item
Partial length Human CDKN2A/p14ARF, expressed in HEK293 cells.
Caution: This product is for Research Use Only. It is not for use in diagnostic or therapeutic procedures.
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:Lyophilised: Store at −20...−80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Gene ID:UniprotID# P42771
- Reconstitution instructions:Prior to reconstitution, centrifuge the vial at 5000 g for 3 - 5 minutes at room temperature. Reconstitute with appropriate volume of distilled sterile water to bring product to 1 mg/ml concentration. After addition of distilled sterile water, mix by gentle tapping. Note: It is not recommended to vortex or vigorously pipette the sample.
- Carrier-free:N
- Protein synonyms:CDK4I|CDKN2|CDKN2A|Cyclin-dependent kinase 4 inhibitor A|Cyclin-dependent kinase inhibitor 2A|MTS-1|MTS1|Multiple tumor suppressor 1|p16-INK4|p16-INK4a
- Protein/peptide name:CDKN2A/p14ARF
- Purity:>95% by SDS-PAGE and Coomassie blue staining
- Molecular weight:The protein has a predicted molecular mass of 29,4 kDa after removal of the signal peptide. The apparent molecular mass of hFc-p16(110-139) is approximately 25 - 35 kDa due to glycosylation.
- Sequence:hFc (Glu99-Ala330) + p16 (Trp110-Gly139)
- Concentration:Reconstitution dependent
- Formulation:Lyophilised from sterile Phosphate Buffered Saline, pH 7,4. Normally 5 - 8% Trehalose is added as a protectant before lyophilization.
- Shipping temperature:Shipped on blue ice at 4 °C
- Tested applications:SDS-PAGE