Human Recombinant PLD4 (from HEK293 Cells)
Supplier: Antibodies.com
New Product
Certificates
About this item
Partial length Human PLD4, expressed in HEK293 cells.
Caution: This product is for esearch use only. It is not for use in diagnostic or therapeutic procedures.
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:Lyophilised: Store at −20...−80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Gene ID:UniprotID# Q96BZ4
- Reconstitution instructions:Reconstitute with distilled sterile water
- Carrier-free:N
- Protein synonyms:5'-3' exonuclease C14orf175|Choline phosphatase 4|Phosphatidylcholine-hydrolyzing phospholipase D4|Phospholipase D family member 4|PLD 4
- Protein/peptide name:PLD4
- Purity:>95% by SDS-PAGE and Coomassie blue staining
- Molecular weight:The protein has a predicted molecular mass of 76,2 kDa after removal of the signal peptide The apparent molecular mass of hFc-PLD4 is approximately 70 - 130 kDa due to glycosylation
- Sequence:hFc (Glu99-Ala330) + PLD4 (Trp52-Gly506)
- Concentration:Reconstitution dependent
- Formulation:Lyophilised from sterile phosphate buffered saline, pH 7,4. Normally 5 - 8% trehalose is added as a protectant before lyophilisation.
- Shipping temperature:Shipped on blue ice at 4 °C
- Tested applications:SDS-PAGE