0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
Proteins and Peptides
Recombinant Human SLPI (from HEK293 cells)
Recombinant Human SLPI (from HEK293 cells)
:
ANTIBODIES.COM
New Product
Recombinant Human SLPI (from HEK293 cells)
:
ANTIBODIES.COM
New Product