Order Entry
Export
ContactUsLinkComponent
 
Human Recombinant sVAP-1 (from HEK293 Cells)
New Product
 
Human Recombinant sVAP-1 (from HEK293 Cells)
New Product

 

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage conditions:
    Upon delivery aliquot and store at −20...−70 °C. Avoid freeze/thaw cycles.
  • Endotoxin content:
    <0,1 ng/µg (1 IEU/µg) as measured by LAL method
  • Gene ID:
    UniprotID# Q16853
  • Carrier-free:
    N
  • Protein/peptide name:
    sVAP-1
  • Purity:
    >95% (by SDS-PAGE)
  • Molecular weight:
    83 kDa (including tags)
  • Sequence:
    HHHHHHHHHHGSGGDGGEPSQLPHCPSVSPSAQPWTHPGQSQLFADLSREELTAVMRFLTQRLGPGLVDAAQARPSDNCVFSVELQLPPKAAALAHLDRGSPPPAREALAIVFFGRQPQPNVSELVVGPLPHPSYMRDVTVERHGGPLPYHRRPVLFQEYLDIDQMIFNRELPQASGLLHHCCFYKHRGRNLVTMTTAPRGLQSGDRATWFGLYYNISGAGFFLHHVGLELLVNHKALDPARWTIQKVFYQGRYYDSLAQLEAQFEAGLVNVVLIPDNGTGGSWSLKSPVPPGPAPPLQFYPQGPRFSVQGSRVASSLWTFSFGLGAFSGPRIFDVRFQGERLVYEISLQEALAIYGGNSPAAMTTRYVDGGFGMGKYTTPLTRGVDCPYLATYVDWHFLLESQAPKTIRDAFCVFEQNQGLPLRRHHSDLYSHYFGGLAETVLVVRSMSTLLNYDYVWDTVFHPSGAIEIRFYATGYISSAFLFGATGKYGNQVSEHTLGTVHTHSAHFKVDLDVAGLENWVWAEDMVFVPMAVPWSPEHQLQRLQVTRKLLEMEEQAAFLVGSATPRYLYLASNHSNKWGHPRGYRIQMLSFAGEPLPQNSSMARGFSWERYQLAVTQRKEEEPSSSSVFNQNDPWAPTVDFSDFINNETIAGKDLVAWVTAGFLHIPHAEDIPNTVTVGNGVGFFLRPYNFFDEDPSFYSADSIYFRGDQDAGACEVNPLACLPQAAACAPDLPAFSHGGFSHN
  • Concentration:
    Lot specific
  • Formulation:
    Supplied in phosphate buffered saline with 20% glycerol
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Tested applications:
    WB, SDS-PAGE, Immunoassays, Functional Studies