Order Entry
Export
ContactUsLinkComponent
 
Human Recombinant IL-17RB (from HEK293 Cells)
New Product
 
Human Recombinant IL-17RB (from HEK293 Cells)
New Product

 

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage conditions:
    Lyophilised: Store at −20...−80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
  • Gene ID:
    UniprotID# Q9NRM6
  • Reconstitution instructions:
    Prior to reconstitution, centrifuge the vial at 5000 g for 3 - 5 minutes at room temperature. Reconstitute with appropriate volume of distilled sterile water to bring product to 1 mg/ml concentration. After addition of distilled sterile water, mix by gentle tapping. Note: It is not recommended to vortex or vigorously pipette the sample.
  • Carrier-free:
    N
  • Protein synonyms:
    CRL4|Cytokine receptor-like 4|EVI27|IL-17 receptor B|IL-17 receptor homolog 1|IL-17B receptor|IL-17Rh1|IL17BR|IL17RB|IL17Rh1
  • Protein/peptide name:
    IL-17RB
  • Purity:
    >95% by SDS-PAGE and Coomassie blue staining
  • Molecular weight:
    The protein has a predicted molecular mass of 55,77 kDa after removal of the signal peptide
  • Sequence:
    IL17RB (Arg18-Pro292) + hFc (Glu99-Ala330)
  • Concentration:
    Reconstitution dependent
  • Formulation:
    Lyophilised from sterile phosphate buffered saline, pH 7,4. Normally 5 - 8% trehalose is added as a protectant before lyophilisation.
  • Shipping temperature:
    Shipped on blue ice at 4 °C