Human Recombinant BCMA (from HEK293 Cells)
Supplier: Antibodies.com
New Product
Certificates
About this item
Partial length Human BCMA, expressed in HEK293 cells.
Caution: This product is for Research Use Only. It is not for use in diagnostic or therapeutic procedures.
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:Lyophilised: Store at −20...−80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Gene ID:UniprotID# Q02223
- Reconstitution instructions:Reconstitute with distilled sterile water
- Carrier-free:N
- Protein synonyms:B-cell maturation protein|BCM|CD269|TNFRSF17|Tumor necrosis factor receptor superfamily member 17
- Protein/peptide name:BCMA
- Purity:>95% by SDS-PAGE and Coomassie blue staining
- Molecular weight:The protein has a predicted molecular mass of 28,6 kDa after removal of the signal peptide. The apparent molecular mass of BCMA(1-22)-mFc is approximately 25 - 35 kDa due to glycosylation.
- Sequence:BCMA (Met1-Ile22) + mFc (Pro99-Lys330)
- Concentration:Reconstitution dependent
- Formulation:Lyophilised from sterile Phosphate Buffered Saline, pH 7,4. Normally 5 - 8% trehalose is added as protectants before lyophilization.
- Shipping temperature:Shipped on blue ice at 4 °C
- Tested applications:SDS-PAGE