Human Recombinant IL-21R (from HEK293 Cells)
New Product
Partial length Human IL-21R, expressed in HEK293 cells, suitable for ELISA, SDS-PAGE.
: This product is for esearch use only. It is not for use in diagnostic or therapeutic procedures.
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:Lyophilised: Store at −20...−80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Gene ID:UniprotID# Q9HBE5
- Reconstitution instructions:Reconstitute with distilled sterile water
- Carrier-free:N
- Protein synonyms:CD360|IL-21 receptor|IL21R|Interleukin-21 receptor|NILR|Novel interleukin receptor
- Protein/peptide name:IL-21R
- Purity:>95% by SDS-PAGE and Coomassie blue staining
- Molecular weight:The protein has a predicted molecular mass of 25,4 kDa after removal of the signal peptide The apparent molecular mass of IL21R-His is approximately 35 - 55 kDa due to glycosylation
- Sequence:IL21R (Cys20-Glu232) + 6×His Tag
- Concentration:Reconstitution dependent
- Formulation:Lyophilised from sterile phosphate buffered saline, pH 7,4. Normally 5 - 8% trehalose is added as a protectant before lyophilisation.
- Shipping temperature:Shipped on blue ice at 4 °C
- Tested applications:ELISA, SDS-PAGE