Order Entry
Export
ContactUsLinkComponent
 
Human Native Human IgG (from Serum)
New Product
 
Human Native Human IgG (from Serum)
New Product

 

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Native
  • Source:
    Serum
  • Species:
    Human
  • Storage conditions:
    Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
  • Carrier-free:
    N
  • Protein synonyms:
    HDC|Heavy chain disease protein|Ig gamma-2 chain C region|Ig gamma-2 chain C region DOT|Ig gamma-2 chain C region TIL|Ig gamma-2 chain C region ZIE|Ig gamma-3 chain C region|Ig gamma-4 chain C region|IGHG2|IGHG3
  • Protein/peptide name:
    Human IgG
  • Purity:
    >95% (SDS-PAGE)
  • Concentration:
    10 mg/ml
  • Formulation:
    Supplied in 0,1 M Phosphate Buffered Saline, pH 7,4, with 0,001% Sodium Azide.
  • Shipping temperature:
    Shipped on blue ice at 4 °C.
  • Tested applications:
    WB, DB, ELISA, SDS-PAGE