Order Entry
Export
ContactUsLinkComponent
Human Recombinant CD22 (from HEK293 Cells)
Human Recombinant CD22 (from HEK293 Cells)
Supplier:  Antibodies.com
New Product
Certificates
Human Recombinant CD22 (from HEK293 Cells)
Supplier:  Antibodies.com
New Product

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage conditions:
    Lyophilised: Store at −20...−80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
  • Gene ID:
    UniprotID# P20273
  • Reconstitution instructions:
    Reconstitute with distilled sterile water
  • Carrier-free:
    N
  • Protein synonyms:
    B-cell receptor CD22|B-lymphocyte cell adhesion molecule|BL-CAM|Sialic acid-binding Ig-like lectin 2|Siglec-2|SIGLEC2|T-cell surface antigen Leu-14
  • Protein/peptide name:
    CD22
  • Purity:
    >95% by SDS-PAGE and Coomassie blue staining
  • Molecular weight:
    The protein has a predicted molecular mass of 55,4 kDa after removal of the signal peptide. The apparent molecular mass of CD22(417-678)-hFc is approximately 70 - 100 kDa due to glycosylation.
  • Sequence:
    CD22 (Tyr417-Tyr678) + hFc (Glu99-Ala330)
  • Concentration:
    Reconstitution dependent
  • Formulation:
    Lyophilised from sterile phosphate buffered saline, pH 7,4. Normally 5 - 8% trehalose is added as protectants before lyophilisation
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Tested applications:
    SDS-PAGE

Specifications