Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Proteins and Peptides
Recombinant Human GPCR GPR75 (from HEK293 cells)
Recombinant Human GPCR GPR75 (from HEK293 cells)
Recombinant Human GPCR GPR75 (from HEK293 cells)
Supplier:
Antibodies.com
New Product
Certificates
Expand image
Recombinant Human GPCR GPR75 (from HEK293 cells)
Supplier:
Antibodies.com
New Product
About this item