Order Entry
Export
ContactUsLinkComponent
 
Human Recombinant MICA (from HEK293 cells)
New Product
 
Human Recombinant MICA (from HEK293 cells)
New Product

 

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage conditions:
    Lyophilised: Store at −20...−80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
  • Gene ID:
    UniprotID# Q29983
  • Reconstitution instructions:
    Reconstitute with distilled sterile water
  • Carrier-free:
    N
  • Protein synonyms:
    MHC class I polypeptide-related sequence A|MIC-A|PERB11.1
  • Protein/peptide name:
    MICA
  • Purity:
    >95% by SDS-PAGE and coomassie blue staining
  • Molecular weight:
    The protein has a predicted molecular mass of 37,9 kDa after removal of the signal peptide. The apparent molecular mass of MICAa3(203-306)-mFc is approximately 35 - 70 kDa due to glycosylation.
  • Sequence:
    MICAa3 (Arg203-His306) + mFc (Pro99-Lys330)
  • Concentration:
    Reconstitution dependent
  • Formulation:
    Lyophilised from sterile Phosphate Buffered Saline, pH 7,4. Normally 5 - 8% trehalose is added as protectants before lyophilisation.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Tested applications:
    SDS-PAGE