Mouse Recombinant CD48 (from HEK293 cells)
New Product
Partial length Mouse CD48, expressed in HEK293 cells.
: This product is for research use only. It is not for use in diagnostic or therapeutic procedures.
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Storage conditions:Lyophilised: Store at −20...−80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Gene ID:UniprotID# P18181
- Reconstitution instructions:Prior to reconstitution, centrifuge the vial at 5000 g for 3 - 5 minutes at room temperature. Reconstitute with appropriate volume of distilled sterile water to bring product to 1 mg/ml concentration. After addition of distilled sterile water, mix by gentle tapping. Note: It is not recommended to vortex or vigorously pipette the sample.
- Carrier-free:N
- Protein synonyms:B-lymphocyte activation marker BLAST-1|BCM1|BCM1 surface antigen|BLAST1|CD48 antigen|Leukocyte antigen MEM-102|Signaling lymphocytic activation molecule 2|SLAM family member 2|SLAMF2|TCT.1
- Protein/peptide name:CD48
- Purity:>95% by SDS-PAGE and coomassie blue staining
- Molecular weight:The protein has a predicted molecular mass of 48,5 kDa after removal of the signal peptide. The apparent molecular mass of mCD48-hFc is approximately 55 - 70 kDa due to glycosylation.
- Sequence:CD48 (Phe23-Ser217) + hFc (Glu99-Ala330)
- Concentration:Reconstitution dependent
- Formulation:Lyophilised from sterile phosphate buffered saline, pH 7,4. Normally 5 - 8% Trehalose is added as a protectant before lyophilisation.
- Shipping temperature:Shipped on blue ice at 4 °C
- Tested applications:SDS-PAGE