Human Recombinant IL-1R-2 (from HEK293 Cells)
New Product
Partial length human IL-1R-2, expressed in HEK293 cells.
: This product is for research use only. It is not for use in diagnostic or therapeutic procedures.
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:Lyophilised: Store at –20...–80 °C. Reconstituted: Aliquot and store at –80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Gene ID:UniprotID# P27930
- Reconstitution instructions:Reconstitute with distilled sterile water
- Carrier-free:N
- Protein synonyms:CD121 antigen-like family member B|CD121b|CDw121b|IL-1 type II receptor|IL-1R-beta|IL-1RT-2|IL-1RT2|IL1R2|IL1RB
- Protein/peptide name:IL-1R-2
- Purity:>85% by SDS-PAGE and Coomassie blue staining
- Molecular weight:The protein has a predicted molecular mass of 38,6 kDa after removal of the signal peptide. The apparent molecular mass of IL1R2-His is approximately 35 - 55 kDa due to glycosylation.
- Sequence:IL1R2 (Phe14-Glu343) + 6×His Tag
- Concentration:Reconstitution dependent
- Formulation:Lyophilised from sterile phosphate buffered saline, pH 7,4. Normally 5 - 8% Trehalose is added as a protectant before lyophilisation.
- Shipping temperature:Shipped on blue ice at 4 °C
- Tested applications:SDS-PAGE