Human Recombinant GDNF (from HEK293 Cells)
New Product
Partial length Human GDNF, expressed in HEK293 cells.
: This product is for research use only. It is not for use in diagnostic or therapeutic procedures.
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:Lyophilised: Store at −20...−80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Gene ID:UniprotID# P39905
- Reconstitution instructions:Prior to reconstitution, centrifuge the vial at 5000g for 3 - 5 minutes at room temperature. Reconstitute with appropriate volume of distilled sterile water to bring product to 1 mg/ml concentration. After addition of distilled sterile water, mix by gentle tapping. Note: It is not recommended to vortex or vigorously pipette the sample.
- Carrier-free:N
- Protein synonyms:Astrocyte-derived trophic factor|ATF|Glial cell line-derived neurotrophic factor|hGDNF
- Protein/peptide name:GDNF
- Purity:>95% by SDS-PAGE and Coomassie blue staining
- Molecular weight:The protein has a predicted molecular mass of 41,2 kDa after removal of the signal peptide. The apparent molecular mass of GDNF-hFc is approximately 55 - 70 kDa due to glycosylation.
- Sequence:GDNF (Ser78-Ile211) + hFc (Glu99-Ala330)
- Concentration:Reconstitution dependent
- Formulation:Lyophilised from sterile phosphate buffered saline, pH 7,4. Normally 5 - 8% trehalose is added as a protectant before lyophilisation.
- Shipping temperature:Shipped on blue ice at 4 °C
- Tested applications:SDS-PAGE