Human Recombinant DC-SIGN (from HEK293 Cells)
Supplier: Antibodies.com
New Product
Certificates
About this item
Partial length Human DC-SIGN, expressed in HEK293 cells.
Caution: This product is for research use only. It is not for use in diagnostic or therapeutic procedures.
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:Lyophilised: Store at −20...−80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Gene ID:UniprotID# Q9NNX6
- Reconstitution instructions:Reconstitute with distilled sterile water
- Carrier-free:N
- Protein synonyms:C-type lectin domain family 4 member L|CD209|CD209 antigen|CLEC4L|DC-SIGN1|Dendritic cell-specific ICAM-3-grabbing non-integrin 1
- Protein/peptide name:DC-SIGN
- Purity:>85% by SDS-PAGE and Coomassie blue staining
- Molecular weight:The protein has a predicted molecular mass of 40,3 kDa after removal of the signal peptide. The apparent molecular mass of His-CD209 is approximately 35 - 55 kDa due to glycosylation.
- Sequence:6×His tag + CD209 (Gln59-Ala404)
- Concentration:Reconstitution dependent
- Formulation:Lyophilised from sterile phosphate buffered saline, pH 7,4. Normally 5 - 8% trehalose is added as protectants before lyophilisation
- Shipping temperature:Shipped on blue ice at 4 °C
- Tested applications:SDS-PAGE