Mouse Recombinant Quiescin Q6 (from HEK293 Cells)
Supplier: Antibodies.com
New Product
Certificates
About this item
Partial length Mouse Quiescin Q6, expressed in HEK293 cells.
Caution: This product is for esearch use only. It is not for use in diagnostic or therapeutic procedures.
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Storage conditions:Lyophilised: Store at −20...−80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Gene ID:UniprotID# Q8BND5
- Reconstitution instructions:Reconstitute with distilled sterile water
- Carrier-free:N
- Protein synonyms:hQSOX|QSCN6|QSOX1|Sulfhydryl oxidase 1
- Protein/peptide name:Quiescin Q6
- Purity:>85% by SDS-PAGE and Coomassie blue staining
- Molecular weight:The protein has a predicted molecular mass of 75,5 kDa after removal of the signal peptide The apparent molecular mass of mQSOX1-His is approximately 70 - 100 kDa due to glycosylation
- Sequence:QSOX1 (Ser36-His Tag708) + 6×His Tag
- Concentration:Reconstitution dependent
- Formulation:Lyophilised from sterile phosphate buffered saline, pH 7,4. Normally 5 - 8% trehalose is added as a protectant before lyophilisation.
- Shipping temperature:Shipped on blue ice at 4 °C
- Tested applications:SDS-PAGE