Human Recombinant TNFSF18/GITRL (from HEK293 Cells)
Supplier: Antibodies.com
New Product
Certificates
About this item
Partial length Human TNFSF18/GITRL, expressed in HEK293 cells, suitable for SDS-PAGE, flow cytometry.
Caution: This product is for esearch use only. It is not for use in diagnostic or therapeutic procedures.
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:Lyophilised: Store at −20...−80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Gene ID:UniprotID# Q9UNG2
- Reconstitution instructions:Reconstitute with distilled sterile water
- Carrier-free:N
- Protein/peptide name:TNFSF18/GITRL
- Purity:>95% by SDS-PAGE and Coomassie blue staining
- Molecular weight:The protein has a predicted molecular mass of 15,3 kDa after removal of the signal peptide
- Sequence:GITR Ligand (Gln50-Ser177) + 6×His Tag
- Concentration:Reconstitution dependent
- Formulation:Lyophilised from sterile phosphate buffered saline, pH 7,4. Normally 5 - 8% trehalose is added as a protectant before lyophilisation.
- Shipping temperature:Shipped on blue ice at 4 °C
- Tested applications:SDS-PAGE, Flow Cytometry