Order Entry
Export
ContactUsLinkComponent
Human Recombinant IL-19 (from HEK293 cells)
Human Recombinant IL-19 (from HEK293 cells)
Supplier:  Antibodies.com
New Product
Certificates
Human Recombinant IL-19 (from HEK293 cells)
Supplier:  Antibodies.com
New Product

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage conditions:
    Lyophilised: Store at −20...−80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
  • Gene ID:
    UniprotID# Q9UHD0
  • Reconstitution instructions:
    Prior to reconstitution, centrifuge the vial at 5000 g for 3 - 5 minutes at room temperature. Reconstitute with appropriate volume of distilled sterile water to bring product to 1 mg/ml concentration. After addition of distilled sterile water, mix by gentle tapping. Note: It is not recommended to vortex or vigorously pipette the sample.
  • Carrier-free:
    N
  • Protein synonyms:
    IL19|Interleukin-19|Melanoma differentiation-associated protein-like protein|NG.1|ZMDA1
  • Protein/peptide name:
    IL-19
  • Purity:
    >95% by SDS-PAGE and coomassie blue staining
  • Molecular weight:
    The protein has a predicted molecular mass of 43,9 kDa after removal of the signal peptide. The apparent molecular mass of IL19-hFc is approximately 35 - 70 kDa due to glycosylation.
  • Sequence:
    IL19 (Leu25-Ala177) + hFc (Glu99-Ala330)
  • Concentration:
    Reconstitution dependent
  • Formulation:
    Lyophilised from sterile phosphate buffered saline, pH 7,4. Normally 5 - 8% Trehalose is added as a protectant before lyophilisation.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Tested applications:
    SDS-PAGE

Specifications