Order Entry
Export
ContactUsLinkComponent
 
Recombinant T4 Gene 32 (from E. coli)
Catalog # ANTIA200-5200
Supplier:  Antibodies.com
New Product
CAS Number:  
Recombinant T4 Gene 32 (from E. coli)
Catalog # ANTIA200-5200
Supplier:  Antibodies.com
New Product
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Seq:
    1442
  • Storage conditions:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Gene ID:
    UniprotID# P03695
  • Carrier-free:
    N
  • Protein/peptide name:
    T4 Gene 32
  • Purity:
    >95% (by SDS-PAGE). The absence of endonucleases and exonucleases was confirmed.
  • Molecular weight:
    33,5 kDa
  • Concentration:
    10 mg/ml
  • Formulation:
    Supplied in 20 mM Tris-HCl, pH 8,0, with 100 mM NaCl, 0,5 mM Dithiothreitol, 1 mM EDTA and 50% Glycerol.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Tested applications:
    SDS-PAGE, Functional Studies

Specifications

About this item

Full length T4 Gene 32, expressed in Escherichia coli, suitable for SDS-PAGE, Functional Studies and is biologically active.

Caution: This product is for research use only. It is not for use in diagnostic or therapeutic procedures.