Mouse Recombinant CCR2 HEK293 Cells,
Supplier: Antibodies.com
New Product
Certificates
About this item
Partial length Mouse CCR2, expressed in HEK293 cells.
Caution: This product is for Research Use Only. It is not for use in diagnostic or therapeutic procedures.
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Storage conditions:Lyophilised: Store at −20...−80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Gene ID:UniprotID# P51683
- Reconstitution instructions:Reconstitute with distilled sterile water
- Carrier-free:N
- Protein synonyms:C-C chemokine receptor type 2|C-C CKR-2|CC-CKR-2|CCR-2|CD192|CMKBR2|MCP-1-R|Monocyte chemoattractant protein 1 receptor
- Protein/peptide name:CCR2
- Purity:>95% by SDS-PAGE and Coomassie blue staining
- Molecular weight:The protein has a predicted molecular mass of 32,3 kDa after removal of the signal peptide. The apparent molecular mass of mCCR2-hFc is approximately 35 - 55 kDa due to glycosylation.
- Sequence:Mouse CCR2 (Met1-Ala55) + hFc (Glu99-Ala330)
- Concentration:Reconstitution dependent
- Formulation:Lyophilised from sterile Phosphate Buffered Saline, pH 7,4. Normally 5 - 8% trehalose is added as protectants before lyophilization.
- Shipping temperature:Shipped on blue ice at 4 °C
- Tested applications:SDS-PAGE