Human Recombinant C6 (from HEK293 Cells)
Supplier: Antibodies.com
New Product
Certificates
About this item
Partial length human C6, expressed in HEK293 cells.
Caution: This product is for research use only. It is not for use in diagnostic or therapeutic procedures.
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:Lyophilised: Store at –20...–80 °C. Reconstituted: Aliquot and store at –80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Gene ID:UniprotID# P13671
- Reconstitution instructions:Reconstitute with distilled sterile water
- Carrier-free:N
- Protein synonyms:Complement component C6
- Protein/peptide name:C6
- Purity:>85% by SDS-PAGE and Coomassie blue staining
- Molecular weight:The protein has a predicted molecular mass of 103,2 kDa after removal of the signal peptide. The apparent molecular mass of C6-His is approximately 100 - 130 kDa due to glycosylation.
- Sequence:C6 (Cys22-Ala934) + 6×His tag
- Concentration:Reconstitution dependent
- Formulation:Lyophilised from sterile phosphate buffered saline, pH 7,4. Normally 5 - 8% trehalose is added as protectants before lyophilisation.
- Shipping temperature:Shipped on blue ice at 4 °C
- Tested applications:SDS-PAGE