Order Entry
Export
ContactUsLinkComponent
Human Recombinant Met (c-Met) (from HEK293 Cells)
Human Recombinant Met (c-Met) (from HEK293 Cells)
Supplier:  Antibodies.com
New Product
Certificates
Human Recombinant Met (c-Met) (from HEK293 Cells)
Supplier:  Antibodies.com
New Product

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage conditions:
    Lyophilised: Store at –20...–80 °C. Reconstituted: Aliquot and store at –80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
  • Gene ID:
    UniprotID# P08581
  • Reconstitution instructions:
    Reconstitute with distilled sterile water.
  • Carrier-free:
    N
  • Protein synonyms:
    Hepatocyte growth factor receptor|HGF receptor|HGF/SF receptor|MET|Proto-oncogene c-Met|Scatter factor receptor|Tyrosine-protein kinase Met
  • Protein/peptide name:
    Met (c-Met)
  • Purity:
    >95% by SDS-PAGE and Coomassie blue staining
  • Molecular weight:
    The protein has a predicted molecular mass of 127,8 kDa after removal of the signal peptide. The apparent molecular mass of MET-hFc is approximately 100 - 250 kDa due to glycosylation.
  • Sequence:
    MET (Glu25-Thr932) + hFc (Glu99-Ala330)
  • Concentration:
    Reconstitution dependent
  • Formulation:
    Lyophilised from sterile Phosphate Buffered Saline, pH 7,4. Normally 5 - 8% trehalose is added as protectants before lyophilisation.
  • Shipping temperature:
    Shipped on blue ice at 4 °C.
  • Tested applications:
    SDS-PAGE

Specifications