
2,4-Dichloro-6-iodoaniline ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:697-90-5
:
Formula:C₆H₄Cl₂IN
MDL Number:MFCD08533279
:287.92 g/mol
Melting Pt:79…81 °C
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
:GHS06