Methyl dimethoxyacetate, Sigma-Aldrich®
Catalog # SIAL294632-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:134.13 g/mol
EINECS:201-950-0
CAS Number:89-91-8
Formula:C₅H₁₀O₄
MDL Number:MFCD00008484