Order Entry
Export
Orders LinkContactUsLinkComponent
 

Chromone, Sigma-Aldrich®

Glass bottle for solids, 5 g
Catalog # SIAL199222-5G
Supplier:   Merck
Chromone, Sigma-Aldrich®
Catalog # SIAL199222-5G
Supplier:  Merck
CAS Number:  491-38-3
Chromone, Sigma-Aldrich®

Glass bottle for solids, 5 g

CAS Number:  491-38-3
Price
81,70 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  4H-Chromen-4-one
  • CAS Number:491-38-3
  • Formula:C₉H₆O₂
  • MDL Number:MFCD00024064
  • Molecular Weight:146.14 g/mol
  •  :M-Clarity™ quality level = MQ100
  • EINECS:207-737-9
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications