3-(2,6-Dichlorophenyl)-5-methylisoxazole-4-carboxamide
Catalog # MAYBCD05279.50
Supplier: Thermo Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:50 g
- Seq:3368
Specifications
About this item
:
CAS Number:23858-59-5
Formula:C₁₁H₈Cl₂N₂O₂
MDL Number:MFCD00052211