EEDQ (ethyl-N-(2-ethoxy-1,2-dihydroquinoline)carboxylate)
Catalog # MOLEM10325546
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:247.29 g/mol
EINECS:240-418-2
CAS Number:16357-59-8
Formula:C₁₄H₁₇NO₃
MDL Number:MFCD00006703