
EEDQ (ethyl-N-(2-ethoxy-1,2-dihydroquinoline)carboxylate) 99+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:16357-59-8
:
Formula:C₁₄H₁₇NO₃
MDL Number:MFCD00006703
Boiling Pt:125…128 °C (.1 mmHg)
Melting Pt:63…66 °C
EINECS:240-418-2
ADR:6.1,III
:247.29 g/mol
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to beige Crystalline powder
- Infrared spectrum:Conforms
- Melting point:63 to 66 °C
- HPLC:≥99.0 %