Order Entry
Export
ContactUsLinkComponent
2,6-Diacetylpyridine 99%
Irritant
2,6-Diacetylpyridine 99%
Certificates
undefined
Irritant
2,6-Diacetylpyridine 99%

Specification Test Results

  • Appearance:
    White to beige Powder or crystals or needles
  • Infrared spectrum:
    Conforms
  • Melting point:
    78 to 82 °C
  • GC:
    ≥98.5 %

Specification Test Results

Related Information