Dimethyl-2,5-pyridinedicarboxylate, Sigma-Aldrich®
Catalog # SIAL540234-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:195.17 g/mol
:M-Clarity™ quality level = MQ100
CAS Number:881-86-7
Formula:C₉H₉NO₄
MDL Number:MFCD00034767