Order Entry
Export
Orders LinkContactUsLinkComponent
2-(4-Chlorophenoxy)nicotinic acid

2-(4-Chlorophenoxy)nicotinic acid

Supplier:   Thermo Scientific
2-(4-Chlorophenoxy)nicotinic acid
Supplier:  Thermo Scientific
Certificates
2-(4-Chlorophenoxy)nicotinic acid

3 Options Available Below

Product Details & Documents
  • CAS Number:51362-37-9
  • Formula:C₁₂H₈ClNO₃
  • MDL Number:MFCD00052116
  • Molecular Weight:249.64 g/mol
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g

  • Glass bottle for solids
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g

  • Glass bottle for solids
    50 g
    Log in for availability
    Log in for availability
    1 * 50 g
    1 * 50 g