Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-(Benzyloxy)-4,5-dimethoxybenzaldehyde
2-(Benzyloxy)-4,5-dimethoxybenzaldehyde
2-(Benzyloxy)-4,5-dimethoxybenzaldehyde
2-(Benzyloxy)-4,5-dimethoxybenzaldehyde
Supplier:
CHEMPUR
Certificates
Expand image
2-(Benzyloxy)-4,5-dimethoxybenzaldehyde
Supplier:
CHEMPUR
About this item
CAS Number:
14382-86-6
:
Formula:
C₁₆H₁₆O₄
MDL Number:
MFCD00183313