Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
20-Hydroxy N-arachidonoyl taurine
20-Hydroxy-N-arachidonoyl taurine
20-Hydroxy-N-arachidonoyl taurine
20-Hydroxy-N-arachidonoyl taurine
Supplier:
Cayman Chemical
Certificates
Expand image
20-Hydroxy-N-arachidonoyl taurine
Supplier:
Cayman Chemical
About this item
:
Formula:
C₂₂H₃₇NO₅S