3,5-Dimethylisoxazole-4-boronic acid
Catalog # MOLEM83808359
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:140.93 g/mol
CAS Number:16114-47-9
Formula:C₅H₈BNO₃
MDL Number:MFCD02677945