1-Methoxy-1,4-cyclohexadiene, Sigma-Aldrich®
Catalog # SIAL216151-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:M-Clarity™ quality level = MQ200
EINECS:220-747-8
CAS Number:2886-59-1
Formula:C₇H₁₀O
MDL Number:MFCD00001536