Order Entry
Export
Orders LinkContactUsLinkComponent
 

6-Bromochromone

Supplier:   CHEMPUR
6-Bromochromone
Supplier:  CHEMPUR
Certificates

2 Options Available Below

Product Details & Documents
Synonyms:  6-Bromo-4H-chromen-4-one
  • CAS Number:51483-92-2
  • Formula:C₉H₅BrO₂
  • MDL Number:MFCD00239369
  • Molecular Weight:225.0388 g/mol
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    250 mg
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg

  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g

This product has been blocked by your organization. Please contact your purchasing department for more information