Order Entry
Export
Orders LinkContactUsLinkComponent
6-Bromochromone

6-Bromochromone

Glass bottle for solids, 5 g
Catalog # COBBOT-0920-5G
Supplier:   COMBI-BLOCKS
6-Bromochromone
Catalog # COBBOT-0920-5G
Supplier:  COMBI-BLOCKS
CAS Number:  51483-92-2
6-Bromochromone

Glass bottle for solids, 5 g

CAS Number:  51483-92-2
Price
Inquire For Price
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  6-Bromo-4H-chromen-4-one
  • CAS Number:51483-92-2
  • Formula:C₉H₅BrO₂
  • MDL Number:MFCD00239369
  • Molecular Weight:225.0388 g/mol
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications